EMAIL THIS PAGE TO A FRIEND
Journal of Cancer Research and Clinical Oncology
Inhibition of 32Dp210 cells harboring T315I mutation by a novel derivative of emodin correlates with down-regulation of BCR-ABL and its downstream signaling pathways.
PMID 25217883