EMAIL THIS PAGE TO A FRIEND
International Immunopharmacology
Endogenous expression pattern of resolvin D1 in a rat model of self-resolution of lipopolysaccharide-induced acute respiratory distress syndrome and inflammation.
PMID 25218162
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09684
Ammonia, puriss., anhydrous, ≥99.9%
H3N