EMAIL THIS PAGE TO A FRIEND
Cancer letters
HMGA2 induces transcription factor Slug expression to promote epithelial-to-mesenchymal transition and contributes to colon cancer progression.
PMID 25218351
Abstract
Epithelial-to-mesenchymal transition (EMT) is considered to play an essential role in progression and metastasis. This study aims to investigate the expression and underlying molecular targets of high-mobility group AT-hook 2 (HMGA2) in the progression of colon cancer. The expression of HMGA2 is upregulated by both active extracellular signal-regulated kinase (ERK)1/2 and TGF-β signaling in colon cancer cells through a series of lentiviral infection and pharmacological assays. HMGA2 knockdown by specific shRNAs attenuates proliferation, motility and invasion of colon cancer cells in vitro and in vivo. Besides, exogenous HMGA2 expression caused EMT in colon cancer cells, which was confirmed by the downregulation of the epithelial markers and the upregulation of the mesenchymal markers. Moreover, HMGA2 positively regulates the Slug expression by directly binding to the regulatory region in Slug promoter. Importantly, the knockdown of Slug could reverse the HMGA2-induced EMT and decrease the migration and invasion ability of colon cancer cells. Taken together, our results reveal a critical role for HMGA2 in promoting EMT, migration, invasion, and proliferation of colon cancer cells, suggesting HMGA2 as a potential molecular target to prevent colon cancer progression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D8417
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC), suitable for fluorescence
C16H15N5 · 2HCl
32670
4′,6-Diamidino-2-phenylindole dihydrochloride, suitable for fluorescence, BioReagent, ≥95.0% (HPLC)
C16H15N5 · 2HCl
GW21186 Anti-CDH1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2501665
Anti-CDH1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB2701861
Anti-cdh1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA004812
Anti-CDH1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2104222
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB4300693
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
SAB2107370
Anti-CDH1 antibody produced in rabbit, affinity isolated antibody
C7855
Monoclonal Anti-Cdh1 antibody produced in mouse, ~2 mg/mL, clone DCS-266, purified immunoglobulin, buffered aqueous solution
WH0000999M1
Monoclonal Anti-CDH1 antibody produced in mouse, clone 3F4, purified immunoglobulin, buffered aqueous solution
AMAB90862
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1170, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90863
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1172, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90865
Monoclonal Anti-CDH1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1180, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700237 Monoclonal Anti-CDH1 antibody produced in mouse, clone 67A4, purified immunoglobulin, buffered aqueous solution