EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Silica-based nanofibers for electrospun ultra-thin layer chromatography.
PMID 25218634
Abstract
Nanofibrous silica-based stationary phases for electrospun ultra-thin layer chromatography (E-UTLC) are described. Nanofibers were produced by electrospinning a solution of silica nanoparticles dispersed in polyvinylpyrrolidone solutions to create composite silica/polymer nanofibers. Stationary phases were created from as-spun nanofibers, or the nanofibers were heated either to crosslink the polyvinylpyrrolidone or to calcine and selectively remove the polymer. As-spun, crosslinked, and calcined nanofibers with similar mat thicknesses (23-25 μm) were evaluated as stationary phases for E-UTLC separations of laser dyes and amino acids and compared to commercial silica TLC plates. As-spun nanofiber plates offered fast mobile phase velocities, but like other polymer-based nanofibers, separations were only compatible with techniques using nonsolvents of the polymer. Crosslinked nanofibers were not as limited in terms of chemical stability, but separations produced tailed spot shapes. No limitations in terms of mobile phases, analyte solvents, and visualization techniques were observed for calcined nanofibers. Highly efficient separations of amino acids were performed in 15 mm on calcined nanofiber plates, with observed plate heights as low as 8.6 μm, and plate numbers as large as 1400. Additional alignment of the nanofibers provided shorter analysis times but also larger spot widths. The extension of stationary phases to silica-based nanofibers vastly expands the range of mobile phases, analyte solvents, and visualization techniques which can be used for E-UTLC separations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19422
1-Butanol, analytical standard
C4H10O
154679
1-Butanol, spectrophotometric grade, 99.5%
C4H10O
19420
1-Butanol, puriss. p.a., ACS reagent, ≥99.5% (GC)
C4H10O
442665
1-Butanol, analytical standard
C4H10O
03-4585
1-Butanol, for HPLC
C4H10O
PHR1209
1-Butanol, Pharmaceutical Secondary Standard; Certified Reference Material
C4H10O
279544
1-Propanol, anhydrous, 99.7%
C3H8O
96566
1-Propanol, analytical standard
C3H8O
09158
1-Propanol, for inorganic trace analysis, ≥99.8%
C3H8O
82092
1-Propanol, purum, ≥99.0% (GC)
C3H8O
W292818
1-Propanol, ≥99%, FG
C3H8O
W292826
1-Propanol, natural, ≥98%, FG
C3H8O
24-6080
1-Propanol, SAJ first grade, ≥99.0%
C3H8O
24-6070
1-Propanol, JIS special grade
C3H8O
1570406
1-Propanol, United States Pharmacopeia (USP) Reference Standard
C3H8O
PHR1208
1-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O
40311
2-Butanone, 5000 μg/mL in methanol: water (9:1), analytical standard
C4H8O
02469
2-Butanone, analytical standard
C4H8O
48877
2-Butanone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
C4H8O
31579
Androsterone, VETRANAL, analytical standard
C19H30O2
A1270000
Arginine, European Pharmacopoeia (EP) Reference Standard
C6H14N4O2
A1330000
Aspartic acid, European Pharmacopoeia (EP) Reference Standard
C4H7NO4
W217808
Butyl alcohol, ≥99.9%, FCC
C4H10O
A9006
DL-Aspartic acid, ≥99% (TLC)
C4H7NO4
78040
DL-Phenylalanine, puriss., ≥99.0% (NT)
C9H11NO2
W372609
DL-Phenylalanine, 99%, FCC
C9H11NO2
147966
DL-Phenylalanine, ReagentPlus®, 99%
C9H11NO2
246654
Heptane, anhydrous, 99%
C7H16
494526
Heptane, biotech. grade, ≥99%
C7H16
154873
Heptane, spectrophotometric grade, 99%
C7H16