EMAIL THIS PAGE TO A FRIEND
Clinical Biochemistry
Circulating osteoprotegerin is increased in the metabolic syndrome and associates with subclinical atherosclerosis and coronary arterial calcification.
PMID 25218813