EMAIL THIS PAGE TO A FRIEND
Experimental neurology
Sucrose consumption test reveals pharmacoresistant depression-associated behavior in two mouse models of temporal lobe epilepsy.
PMID 25220610
Abstract
Among the comorbidities observed in epilepsy patients depression is the most frequent one. Likewise, depression by itself is accompanied by an increased risk to develop epilepsy. Both epilepsy and depression are characterized by a high incidence of pharmacoresistance, which might be based on overactivity of multidrug transporters like P-glycoprotein at the blood-brain barrier. Using genetically modified mice in preclinical epilepsy research is pivotal for investigating this bidirectional relationship. In the present study, we used the sucrose consumption test (SCT) in the pilocarpine and the intrahippocampal kainate mouse post-status epilepticus model to reveal anhedonic behavior, i.e. hyposensitivity to pleasure, as a key symptom of depression. Mice were repetitively investigated by SCT during early epilepsy and the chronic phase of the disease, during which response to antidepressant drug treatment was assessed. SCT revealed long-lasting anhedonia in both models. Anhedonia appeared to be pharmacoresistant, as neither chronic treatment with imipramine in the pilocarpine model nor chronic treatment with fluoxetine in the kainate model could annihilate the differences in sucrose consumption between control and epileptic mice. Moreover, knock-out of P-glycoprotein did not improve the treatment effect of fluoxetine. In conclusion, our findings show for the first time that the SCT is suited for detection of depression-like behavior in mouse models of temporal-lobe epilepsy. Both models might serve as tools to further investigate the neurobiology and pharmacology of epilepsy-associated pharmacoresistant depression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
15307
Chloral hydrate, meets analytical specification of Ph. Eur., BP, USP, 99.5-101%
C2H3Cl3O2
C8383
Chloral hydrate
C2H3Cl3O2
F132
Fluoxetine hydrochloride, solid
C17H18F3NO · HCl
1279804
Fluoxetine hydrochloride, United States Pharmacopeia (USP) Reference Standard
C17H18F3NO · HCl
F0253000
Fluoxetine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C17H18F3NO · HCl
34012
Fluoxetine hydrochloride, VETRANAL®, analytical standard
C17H18F3NO · HCl
PHR1394
Fluoxetine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C17H18F3NO · HCl
Y0000893
Imipramine for system suitability, European Pharmacopoeia (EP) Reference Standard
C19H24N2 · HCl
I7379
Imipramine hydrochloride, ≥99% (TLC)
C19H24N2 · HCl
I0899
Imipramine hydrochloride, BioXtra, ≥99% (TLC)
C19H24N2 · HCl
1338007
Imipramine hydrochloride, United States Pharmacopeia (USP) Reference Standard
C19H24N2 · HCl
I0100000
Imipramine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C19H24N2 · HCl
I-902
Imipramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
C19H24N2 · HCl
41131
Mettler-Toledo Calibration substance ME 51143091, Saccharin, traceable to primary standards (LGC)
C7H5NO3S
P6503
Pilocarpine hydrochloride, ≥99% (titration), powder
C11H16N2O2 · HCl
P0472
Pilocarpine hydrochloride, meets USP testing specifications
C11H16N2O2 · HCl
P1650000
Pilocarpine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C11H16N2O2 · HCl
240931
Saccharin, ≥99%
C7H5NO3S
109185
Saccharin, ≥98%
C7H5NO3S
PHR1341
Saccharin, Pharmaceutical Secondary Standard; Certified Reference Material
C7H5NO3S
1607007
Saccharin, United States Pharmacopeia (USP) Reference Standard
C7H5NO3S
S0040000
Saccharin, European Pharmacopoeia (EP) Reference Standard
C7H5NO3S