EMAIL THIS PAGE TO A FRIEND
International journal of molecular sciences
Identification of apolipoprotein C-I as a potential Wilms' tumor marker after excluding inflammatory factors.
PMID 25222555
Abstract
Wilms' tumor is one of the most common malignant tumors observed in children, and its early diagnosis is important for late-stage treatment and prognosis. We previously screened and identified protein markers for Wilms' tumor; however, these markers lacked specificity, and some were associated with inflammation. In the current study, serum samples from children with Wilms' tumors were compared with those of healthy controls and patients with systemic inflammatory response syndrome (SIRS). After exclusion of factors associated with inflammation, specific protein markers for Wilms' tumors were identified. After comparing the protein peak values obtained from all three groups, a protein with a m/z of 6438 Da was specified. Purification and identification of the target protein using high-pressure liquid chromatography (HPLC) and two-dimensional liquid chromatography-linearion trap mass spectrometry(2D-LC-LTQ-MS) mass spectrometry, respectively, revealed that it was apolipoprotein C-I (APO C-I). Thus, APO C-I is a specific protein marker for Wilms' tumor.