EMAIL THIS PAGE TO A FRIEND
Journal of neuroscience research
Neuroanatomical circuitry mediating the sensory impact of nicotine in the central nervous system.
PMID 25223294
Abstract
Direct actions of nicotine in the CNS appear to be essential for its reinforcing properties. However, activation of nicotinic acetylcholine receptors (nAChRs) on afferent sensory nerve fibers is an important component of addiction to, and withdrawal from, cigarette smoking. The aim of the present study was to identify the neuroanatomical substrates activated by the peripheral actions of nicotine and to determine whether these sites overlap brain structures stimulated by direct actions of nicotine. Mouse brains were examined by immunohistochemistry for c-Fos protein after intraperitoneal injection of either nicotine hydrogen tartrate salt (NIC; 30 and 40 μg/kg) or nicotine pyrrolidine methiodide (NIC-PM; 20 and 30 μg/kg). NIC-PM induced c-Fos immunoreactivity (IR) at multiple brain sites. In the brainstem, c-Fos IR was detected in the locus coeruleus, laterodorsal tegmental nucleus, and pedunculotegmental nucleus. In the midbrain, c-Fos IR was observed in areas overlapping the ventral tegmental area (VTA), which includes the paranigral nucleus, parainterfascicular nucleus, parabrachial pigmental area, and rostral VTA. Other structures of the nicotine brain-reward circuitry activated by NIC-PM included the hypothalamus, paraventricular thalamic nucleus, lateral habenular nucleus, hippocampus, amygdala, accumbens nucleus, piriform cortex, angular insular cortex, anterior olfactory nucleus, lateral septal nucleus, bed nucleus of stria terminalis, cingulate and medial prefrontal cortex, olfactory tubercle, and medial and lateral orbital cortex. NIC, acting through central and peripheral nAChRs, produced c-Fos IR in areas that overlapped NIC-PM-induced c-Fos-expressing sites. These neuroanatomical data are the first to demonstrate that the CNS structures that are the direct targets of nicotine are also anatomical substrates for the peripheral sensory impact of nicotine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1236
(−)-Nicotine hydrogen tartrate salt, ≥98% (HPLC)
C10H14N2 · 2C4H6O6