EMAIL THIS PAGE TO A FRIEND
Annals of oncology : official journal of the European Society for Medical Oncology
Survival after neoadjuvant chemotherapy with or without bevacizumab or everolimus for HER2-negative primary breast cancer (GBG 44-GeparQuinto)†.
PMID 25223482
Abstract
The GeparQuinto study showed that adding bevacizumab to 24 weeks of anthracycline-taxane-based neoadjuvant chemotherapy increases pathological complete response (pCR) rates overall and specifically in patients with triple-negative breast cancer (TNBC). No difference in pCR rate was observed for adding everolimus to paclitaxel in nonearly responding patients. Here, we present disease-free (DFS) and overall survival (OS) analyses. Patients (n = 1948) with HER2-negative tumors of a median tumor size of 4 cm were randomly assigned to neoadjuvant treatment with epirubicin/cyclophosphamide followed by docetaxel (EC-T) with or without eight infusions of bevacizumab every 3 weeks before surgery. Patients without clinical response to EC ± Bevacizumab were randomized to 12 weekly cycles paclitaxel with or without everolimus 5 mg/day. To detect a hazard ratio (HR) of 0.75 (α = 0.05, β = 0.8) 379 events had to be observed in the bevacizumab arms. With a median follow-up of 3.8 years, 3-year DFS was 80.8% and 3-year OS was 89.7%. Outcome was not different for patients receiving bevacizumab (HR 1.03; P = 0.784 for DFS and HR 0.974; P = 0.842 for OS) compared with patients receiving chemotherapy alone. Patients with TNBC similarly showed no improvement in DFS (HR = 0.99; P = 0.941) and OS (HR = 1.02; P = 0.891) when treated with bevacizumab. No other predefined subgroup (HR+/HER2-; locally advanced (cT4 or cN3) or not; cT1-3 or cT4; pCR or not) showed a significant benefit. No difference in DFS (HR 0.997; P = 0.987) and OS (HR 1.11; P = 0.658) was observed for nonearly responding patients receiving paclitaxel with or without everolimus overall as well as in subgroups. Long-term results, in opposite to the results of pCR, do not support the neoadjuvant use of bevacizumab in addition to an anthracycline-taxane-based chemotherapy or everolimus in addition to paclitaxel for nonearly responding patients. NCT 00567554, www.clinicaltrials.gov.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3250000
Cyclophosphamide, European Pharmacopoeia (EP) Reference Standard
C7H15Cl2N2O2P · H2O