EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
Chemometrics-enhanced high performance liquid chromatography-diode array detection strategy for simultaneous determination of eight co-eluted compounds in ten kinds of Chinese teas using second-order calibration method based on alternating trilinear decomposition algorithm.
PMID 25223614