EMAIL THIS PAGE TO A FRIEND
Journal of proteome research
Quantitative phosphoproteomic analysis of signaling downstream of the prostaglandin e2/g-protein coupled receptor in human synovial fibroblasts: potential antifibrotic networks.
PMID 25223752
Abstract
The Prostaglandin E2 (PGE2) signaling mechanism within fibroblasts is of growing interest as it has been shown to prevent numerous fibrotic features of fibroblast activation with limited evidence of downstream pathways. To understand the mechanisms of fibroblasts producing tremendous amounts of PGE2 with autocrine effects, we apply a strategy of combining a wide-screening of PGE2-induced kinases with quantitative phosphoproteomics. Our large-scale proteomic approach identified a PKA signal transmitted through phosphorylation of its substrates harboring the R(R/X)X(S*/T*) motif. We documented 115 substrates, of which 72 had 89 sites with a 2.5-fold phosphorylation difference in PGE2-treated cells than in untreated cells, where approximately half of such sites were defined as being novel. They were compiled by networking software to focus on highlighted activities and to associate them with a functional readout of fibroblasts. The substrates were associated with a variety of cellular functions including cytoskeletal structures (migration/motility), regulators of G-protein coupled receptor function, protein kinases, and transcriptional/translational regulators. For the first time, we extended the PGE2 pathway into an elaborate network of interconnecting phosphoproteins, providing vital information to a once restricted signalosome. These data provide new insights into eicosanoid-initiated cell signaling with regards to the regulation of fibroblast activation and the identification of new targets for evidenced-based pharmacotherapy against fibrosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
01700
Acrylamide, purum, ≥98.0% (GC)
C3H5NO
01696
Acrylamide, for Northern and Southern blotting, powder blend
C3H5NO
08267
Acrylamide, certified reference material, TraceCERT®
C3H5NO
23701
Acrylamide, analytical standard
C3H5NO
01697
Acrylamide solution, 40% in H2O, for molecular biology
C3H5NO
A9415
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
C62H86N12O16
A4262
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
C62H86N12O16
A3678
Ammonium persulfate, for molecular biology, for electrophoresis, ≥98%
H8N2O8S2
A7460
Ammonium persulfate, BioXtra, ≥98.0%
H8N2O8S2
09913
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
H8N2O8S2
31117
Ammonium persulfate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
H8N2O8S2
02487
Chloroform, analytical standard
CHCl3
PHR1552
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
CHCl3
613339
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
CH3NO
551384
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
CH3NO
612707
Formamide solution, NMR reference standard, 45% in DMSO-d6 (99.9 atom % D), NMR tube size 3 mm × 8 in.
CH3NO
611891
Formamide solution, NMR reference standard, 45% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
CH3NO
733784
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
CH3NO
736201
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
CH3NO
146072
N,N′-Methylenebis(acrylamide), 99%
C7H10N2O2
M7279
N,N′-Methylenebisacrylamide, powder, for molecular biology, for electrophoresis, ≥99.5%
C7H10N2O2
M7256
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
C7H10N2O2
M1533
N,N′-Methylenebisacrylamide solution, for electrophoresis, 2% in H2O
C7H10N2O2
T9281
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ~99%
C6H16N2
T7024
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, for molecular biology, ≥99% (GC)
C6H16N2
411019
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
C6H16N2
T22500
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
C6H16N2
PHR1316
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
CHCl3
S8692
Sulprostone, ≥95% (HPLC), oil
C23H31NO7S