EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
The neuroprotective properties of the superoxide dismutase mimetic tempol correlate with its ability to reduce pathological glutamate release in a rodent model of stroke.
PMID 25224033
Abstract
The contribution of oxidative stress to ischemic brain damage is well established. Nevertheless, for unknown reasons, several clinically tested antioxidant therapies have failed to show benefits in human stroke. Based on our previous in vitro work, we hypothesized that the neuroprotective potency of antioxidants is related to their ability to limit the release of the excitotoxic amino acids glutamate and aspartate. We explored the effects of two antioxidants, tempol and edaravone, on amino acid release in the brain cortex, in a rat model of transient occlusion of the middle cerebral artery (MCAo). Amino acid levels were quantified using a microdialysis approach, with the probe positioned in the ischemic penumbra as verified by a laser Doppler technique. Two-hour MCAo triggered a dramatic increase in the levels of glutamate, aspartate, taurine, and alanine. Microdialysate delivery of 10mM tempol reduced the amino acid release by 60-80%, whereas matching levels of edaravone had no effect. In line with these data, an intracerebroventricular injection of tempol but not edaravone (500 nmol each, 15 min before MCAo) reduced infarction volumes by ~50% and improved neurobehavioral outcomes. In vitro assays showed that tempol was superior at removing superoxide anion, whereas edaravone was more potent at scavenging hydrogen peroxide, hydroxyl radical, and peroxynitrite. Overall, our data suggest that the neuroprotective properties of tempol are probably related to its ability to reduce tissue levels of the superoxide anion and pathological glutamate release and, in such a way, limit progression of brain infarction within ischemic penumbra. These new findings may be instrumental in developing new antioxidant therapies for treatment of stroke.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1378
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
C15H24O
34750
2,6-Di-tert-butyl-4-methylphenol, purum, ≥99.0% (GC)
C15H24O
03762
2,6-Di-tert-butyl-4-methylphenol, tested according to Ph. Eur.
C15H24O
M70800
3-Methyl-1-phenyl-2-pyrazoline-5-one, 99%
C10H10N2O
176141
4-Hydroxy-TEMPO, 97%
C9H18NO2
W218405
Butylated hydroxytoluene, ≥99%, FCC, FG
C15H24O
1082708
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
C15H24O
PHR1117
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
C15H24O
B103500
Butyric acid, ≥99%
C4H8O2
W222119
Butyric acid, natural, ≥99%, FCC, FG
C4H8O2
19215
Butyric acid, analytical standard
C4H8O2
W222100
Butyric acid, ≥99%, FG
C4H8O2
Y0000441
Didanosine impurity A,, European Pharmacopoeia (EP) Reference Standard
C5H4N4O
H9377
Hypoxanthine, ≥99.0%
C5H4N4O
H9636
Hypoxanthine, powder, BioReagent, suitable for cell culture
C5H4N4O
77662
Hypoxanthine, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
C5H4N4O
M8010
N,N′-Dimethyl-9,9′-biacridinium dinitrate, used as chemiluminescent reagent
C28H22N4O6
Y0001138
Phenazone impurity A, European Pharmacopoeia (EP) Reference Standard
C10H10N2O
244864
Potassium, chunks (in mineral oil), 98% trace metals basis
K
679909
Potassium, cubes (in mineral oil), 99.5% trace metals basis
K
244856
Potassium, ingot, 99.95% trace metals basis
K
215813
Potassium hydride, 30 wt % dispersion in mineral oil
HK
708860
Potassium hydride, in paraffin
HK
X7375
Xanthine, ≥99%
C5H4N4O2
X0626
Xanthine, ≥99.5% (HPLC), purified by recrystallization
C5H4N4O2
X4002
Xanthine, BioUltra, ≥99%
C5H4N4O2