EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Reversible covalent inhibition of eEF-2K by carbonitriles.
PMID 25224652
Abstract
eEF-2K is a potential target for treating cancer. However, potent specific inhibitors for this enzyme are lacking. Previously, we identified 2,6-diamino-4-(2-fluorophenyl)-4H-thiopyran-3,5-dicarbonitrile (DFTD) as an inhibitor of eEF-2K. Here we describe its mechanism of action against eEF-2K, on the basis of kinetic, mutational, and docking studies, and use chemoinformatic approaches to identify a similar class of carbonitrile-containing compounds that exhibit the same mechanism of action. We show that DFTD behaves as a reversible covalent inhibitor of eEF-2K with a two-step mechanism of inhibition: a fast initial binding step, followed by a slower reversible inactivation step. Molecular docking suggests that a nitrile group of DFTD binds within 4.5 Å of the active site Cys146 to form a reversible thioimidate adduct. Because Cys146 is not conserved amongst other related kinases, targeting this residue holds promise for the development of selective covalent inhibitors of eEF-2K.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00990
Acetyl chloride, puriss. p.a., ≥99.0% (T)
C2H3ClO
434760
Benzamidine hydrochloride, 99%
C7H8N2 · HCl