EMAIL THIS PAGE TO A FRIEND
Journal of Radiation Research
Combination of macrophage inflammatory protein 1 alpha with existing therapies to enhance the antitumor effects on murine hepatoma.
PMID 25225286
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46209
Picric acid solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
C6H3N3O7