EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Mucosal-associated invariant T cell deficiency in systemic lupus erythematosus.
PMID 25225673
Abstract
Mucosal-associated invariant T (MAIT) cells contribute to protection against certain microorganism infections and play an important role in mucosal immunity. However, the role of MAIT cells remains enigmatic in autoimmune diseases. In this study, we examined the level and function of MAIT cells in patients with rheumatic diseases. MAIT cell, cytokine, and programmed death-1 (PD-1) levels were measured by flow cytometry. Circulating MAIT cell levels were significantly reduced in systemic lupus erythematosus (SLE) and rheumatoid arthritis patients. In particular, this MAIT cell deficiency was more prominent in CD8(+) and double-negative T cell subsets, and significantly correlated with disease activity, such as SLE disease activity index and 28-joint disease activity score. Interestingly, MAIT cell frequency was significantly correlated with NKT cell frequency in SLE patients. IFN-γ production in MAIT cells was impaired in SLE patients, which was due to an intrinsic defect in the Ca(2+)/calcineurin/NFAT1 signaling pathway. In SLE patients, MAIT cells were poorly activated by α-galactosylceramide-stimulated NKT cells, thereby showing the dysfunction between MAIT cells and NKT cells. Notably, an elevated expression of PD-1 in MAIT cells and NKT cells was associated with SLE. In rheumatoid arthritis patients, MAIT cell levels were significantly higher in synovial fluid than in peripheral blood. Our study primarily demonstrates that MAIT cells are numerically and functionally deficient in SLE. In addition, we report a novel finding that this MAIT cell deficiency is associated with NKT cell deficiency and elevated PD-1 expression. These abnormalities possibly contribute to dysregulated mucosal immunity in SLE.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4700041 Anti-CD3, low endotoxin antibody, Mouse monoclonal, clone OKT3, purified immunoglobulin, buffered aqueous solution
SAB4300686
Anti-CD3E antibody produced in rabbit, affinity isolated antibody
HPA039113
Anti-KLRB1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
30024
Cyclosporin A, 98.5-101.5% (on dried basis)
C62H111N11O12
C3662
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
C62H111N11O12
C1832
Cyclosporin A, from Tolypocladium inflatum, BioReagent, for molecular biology, ≥95%
C62H111N11O12
32425
Cyclosporin A, VETRANAL®, analytical standard
C62H111N11O12
PHR1092
Cyclosporine, Pharmaceutical Secondary Standard; Certified Reference Material
C62H111N11O12
SAB4700040 Monoclonal Anti-CD3 antibody produced in mouse, clone OKT3, purified immunoglobulin, buffered aqueous solution
SAB4700050 Monoclonal Anti-CD3 antibody produced in mouse, clone APA1/1, purified immunoglobulin, buffered aqueous solution
SAB4700044 Monoclonal Anti-CD3 antibody produced in mouse, clone MEM-57, purified immunoglobulin, buffered aqueous solution
SAB4700045 Monoclonal Anti-CD3 antibody produced in mouse, clone MEM-92, purified immunoglobulin, buffered aqueous solution
C7048
Monoclonal Anti-CD3 antibody produced in mouse, clone UCHT-1, purified immunoglobulin, buffered aqueous solution
WH0000916M4
Monoclonal Anti-CD3E antibody produced in mouse, clone 4C1, purified immunoglobulin, buffered aqueous solution
AMAB90876
Monoclonal Anti-CD3E antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1466, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90879
Monoclonal Anti-CD3E antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1497, purified immunoglobulin, buffered aqueous glycerol solution
WH0003820M1
Monoclonal Anti-KLRB1 antibody produced in mouse, clone 2F3, purified immunoglobulin, buffered aqueous solution