EMAIL THIS PAGE TO A FRIEND
Archives of Environmental Contamination and Toxicology
A candidate short-term toxicity test using Ampelisca brevicornis to assess sublethal responses to pharmaceuticals bound to marine sediments.
PMID 25227176