EMAIL THIS PAGE TO A FRIEND
BioMed Research International
Anticancer drug 2-methoxyestradiol protects against renal ischemia/reperfusion injury by reducing inflammatory cytokines expression.
PMID 25229058