EMAIL THIS PAGE TO A FRIEND
Nature
Artificial sweeteners induce glucose intolerance by altering the gut microbiota.
PMID 25231862
Abstract
Non-caloric artificial sweeteners (NAS) are among the most widely used food additives worldwide, regularly consumed by lean and obese individuals alike. NAS consumption is considered safe and beneficial owing to their low caloric content, yet supporting scientific data remain sparse and controversial. Here we demonstrate that consumption of commonly used NAS formulations drives the development of glucose intolerance through induction of compositional and functional alterations to the intestinal microbiota. These NAS-mediated deleterious metabolic effects are abrogated by antibiotic treatment, and are fully transferrable to germ-free mice upon faecal transplantation of microbiota configurations from NAS-consuming mice, or of microbiota anaerobically incubated in the presence of NAS. We identify NAS-altered microbial metabolic pathways that are linked to host susceptibility to metabolic disease, and demonstrate similar NAS-induced dysbiosis and glucose intolerance in healthy human subjects. Collectively, our results link NAS consumption, dysbiosis and metabolic abnormalities, thereby calling for a reassessment of massive NAS usage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5139
Asp-Phe methyl ester, ≥98%
C14H18N2O5
47135
Aspartame, analytical standard
C14H18N2O5
1043706
Aspartame, United States Pharmacopeia (USP) Reference Standard
C14H18N2O5
A1320000
Aspartame, European Pharmacopoeia (EP) Reference Standard
C14H18N2O5
PHR1381
Aspartame, Pharmaceutical Secondary Standard; Certified Reference Material
C14H18N2O5
17850
Ciprofloxacin, ≥98.0% (HPLC)
C17H18FN3O3
M1547
Metronidazole, BioXtra
C6H9N3O3
41131
Mettler-Toledo Calibration substance ME 51143091, Saccharin, traceable to primary standards (LGC)
C7H5NO3S
240931
Saccharin, ≥99%
C7H5NO3S
1607007
Saccharin, United States Pharmacopeia (USP) Reference Standard
C7H5NO3S
PHR1341
Saccharin, Pharmaceutical Secondary Standard; Certified Reference Material
C7H5NO3S
S0040000
Saccharin, European Pharmacopoeia (EP) Reference Standard
C7H5NO3S
B5887
Sodium butyrate, ≥98.5% (GC)
C4H7NaO2
P1880
Sodium propionate, ≥99.0%
C3H5NaO2
69293
Sucralose, ≥98.0% (HPLC)
C12H19Cl3O8
90984
Sucralose, analytical standard
C12H19Cl3O8
1623626
Sucralose, United States Pharmacopeia (USP) Reference Standard
C12H19Cl3O8
PHR1342
Sucralose, Pharmaceutical Secondary Standard; Certified Reference Material
C12H19Cl3O8
Y0001344
Sucralose, European Pharmacopoeia (EP) Reference Standard
C12H19Cl3O8
S7903
Sucrose, BioXtra, ≥99.5% (GC)
C12H22O11
SBR00001
Vancomycin hydrochloride, Ready Made Solution, 100 mg/mL in DMSO, 0.2 μm filtered
C66H75Cl2N9O24 · xHCl