EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Selective filtering defect at the axon initial segment in Alzheimer's disease mouse models.
PMID 25232037
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU093441 MISSION® esiRNA, esiRNA human ANK3 (esiRNA1)