EMAIL THIS PAGE TO A FRIEND
Nanoscale Research Letters
In vitro performance of lipid-PLGA hybrid nanoparticles as an antigen delivery system: lipid composition matters.
PMID 25232295
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6354
1,2-Dioleoyl-sn-glycero-3-phosphocholine, lyophilized powder
C44H84NO8P
561932
Deblock trichloroacetic acid solution, 3 % (w/v) in methylene chloride
C2HCl3O2
270997
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
CH2Cl2
443484
Dichloromethane, ACS reagent, ≥99.5%, contains 50 ppm amylene as stabilizer
CH2Cl2
439223
Dichloromethane, for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
CH2Cl2
270563
Dichloromethane, ≥99.9%
CH2Cl2
66742
Dichloromethane, Selectophore, ≥99.5%
CH2Cl2
02575
Dichloromethane, analytical standard
CH2Cl2
48538
Dichloromethane, analytical standard, stabilized
CH2Cl2
07-2205
Dichloromethane, suitable for PCB analysis
CH2Cl2
07-2206
Dichloromethane, JIS 5000, for residue analysis
CH2Cl2
07-2230
Dichloromethane, for HPLC
CH2Cl2
48600
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
CH2Cl2
40042
Dichloromethane solution, certified reference material, 5000 μg/mL in methanol
CH2Cl2
723851
Dichloromethane solution, contains 10 % (v/v) methanol
CH2Cl2
723878
Dichloromethane solution, 10 % (v/v) in methanol, 1 % (v/v) in ammonium hydroxide
CH2Cl2
03609
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
C10H16N2O8
03620
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
C10H16N2O8
03690
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
C10H16N2O8
M1550000
Methylene chloride, European Pharmacopoeia (EP) Reference Standard
CH2Cl2
PHR1557
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
CH2Cl2
R6626
Rhodamine B, ≥95% (HPLC)
C28H31ClN2O3
83689
Rhodamine B, for fluorescence
C28H31ClN2O3
79754
Rhodamine B, analytical standard
C28H31ClN2O3
02558
Rhodamine B solution, 0.2% in isopropanol, for TLC derivatization
C28H31ClN2O3
S1827
Sodium deoxycholate, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production
C24H39NaO4
71726
Sodium dodecyl sulfate, for ion pair chromatography, ≥99.0%
C12H25NaO4S
71727
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
C12H25NaO4S
Y0000620
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
C12H25NaO4S
672920
TCA Deblock, 3 % (w/v) in methylene chloride
C2HCl3O2