EMAIL THIS PAGE TO A FRIEND
BMJ open
The expression of chemokine receptors CXCR3 and CXCR4 in predicting postoperative tumour progression in stages I-II colon cancer: a retrospective study.
PMID 25232565
Abstract
The prognostic significance of chemokine receptors in stage I/II colon cancer is unclear. We assessed the prognostic value of chemokine receptor CXCR3 and CXCR4 in stage I/II colon cancer. 145 patients with stage I/II colon cancer who underwent curative surgery alone from 2000 to 2007 were investigated. Chemokine receptor expression was assessed by immunohistochemistry. The associations between CXCR3, CXCR4 and clinicopathological variables were analysed using the χ2 test, and the relationships between chemokine receptors and a 5-year disease-free survival were analysed by univariate and multivariate analyses. The high-expression rates of CXCR3 and CXCR4 were 17.9% (26/145) and 38.6% (56/145), respectively. There were no significant associations between the expressions of CXCR3, CXCR4 and clinicopathological factors including gender, age, tumour location, histological differentiation, pathological stage, lymphovascular invasion and pretreatment serum carcinoembryonic antigen (CEA). The 5-year disease-free survival was not significantly different between low-expression groups and high-expression groups of CXCR3 and CXCR4. Multivariate analysis revealed that serum CEA and a number of retrieved lymph nodes, rather than chemokine receptors, were independent prognosticators. CXCR3 and CXCR4 are not independent prognosticators for stage I/II colon cancer after curative surgery.