EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Synthesis of tetra-substituted imidazoles and 2-imidazolines by Ni(0)-catalyzed dehydrogenation of benzylic-type imines.
PMID 25232889
Abstract
Ni(0)-catalyzed dehydrogenation of benzylic-type imines was performed to yield asymmetrical tetra-substituted imidazoles and 2-imidazolines. This was achieved with a single operational step while maintaining good selectivity and atom economy. The catalytic system shows low to moderate tolerance for fluoro-, trifluoromethyl-, methyl-, and methoxy-substituted benzylic-type imines. In addition, the substitution pattern at the N-heterocyclic products was easily controlled by the appropriate selection of R-groups in the starting organic substrates. Based on experimental observations, we propose a reaction mechanism in which benzylic C(sp(3))-H bond activation and insertion steps play pivotal roles in this nickel-catalyzed organic transformation.