EMAIL THIS PAGE TO A FRIEND
JAMA ophthalmology
Trypan blue vital dye staining vs TUNEL technique to detect corneal endothelium toxic effects.
PMID 25233280
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9819
Chondroitin sulfate A sodium salt from bovine trachea, lyophilized powder, BioReagent, suitable for cell culture
C4384 Chondroitin sulfate sodium salt from shark cartilage, sulfated glycosaminoglycan polysaccharide
00268 Dextran, analytical standard, for GPC, 1,000
00269 Dextran, analytical standard, for GPC, 5,000
00270 Dextran, analytical standard, for GPC, 12,000
00271 Dextran, analytical standard, for GPC, 25,000
00891 Dextran, analytical standard, for GPC, 50,000
00892 Dextran, analytical standard, for GPC, 80,000
00893 Dextran, analytical standard, for GPC, 150,000
00894 Dextran, analytical standard, for GPC, 270,000
00895 Dextran, analytical standard, for GPC, 410,000
00896 Dextran, analytical standard, for GPC, 670,000
31394 Dextran, enzymatic synth.
31430 Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
D0731000 Dextran, European Pharmacopoeia (EP) Reference Standard
D0731005 Dextran 1, European Pharmacopoeia (EP) Reference Standard
D0734000 Dextran 10 for calibration, European Pharmacopoeia (EP) Reference Standard
D0737000 Dextran 250 for calibration, European Pharmacopoeia (EP) Reference Standard
D0733000 Dextran 4 for calibration, European Pharmacopoeia (EP) Reference Standard
D0735000 Dextran 40 for calibration, European Pharmacopoeia (EP) Reference Standard
D0738000 Dextran 40 for performance test, European Pharmacopoeia (EP) Reference Standard
D0739000 Dextran 60/70 for performance test, European Pharmacopoeia (EP) Reference Standard
D0736000 Dextran 70 for calibration, European Pharmacopoeia (EP) Reference Standard
31416 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 1,000
31417 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 5,000
D9260 Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
31418 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 12,000
D4626 Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
31419 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 25,000
D1662 Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000