EMAIL THIS PAGE TO A FRIEND
Experimental and Molecular Pathology
Analysis of biological and technical variability in gene expression assays from formalin-fixed paraffin-embedded classical Hodgkin lymphomas.
PMID 25236575