EMAIL THIS PAGE TO A FRIEND
Experimental and molecular pathology
Heme oxygenase-1 has antitumoral effects in colorectal cancer: involvement of p53.
PMID 25236576
Abstract
The expression of heme oxygenase-1 (HO-1) has been shown to be up-regulated in colorectal cancer (CRC), but the role it plays in this cancer type has not yet been addressed. The aims of this study have been to analyze HO-1 expression in human invasive CRC, evaluate its correlation with clinical and histo-pathological parameters and to investigate the mechanisms through which the enzyme influences tumor progression. We confirmed that HO-1 was over-expressed in human invasive CRC and found that the expression of the enzyme was associated with a longer overall survival time. In addition, we observed in a chemically-induced CRC animal model that total and nuclear HO-1 expression increases with tumor progression. Our investigation of the mechanisms involved in HO-1 action in CRC demonstrates that the protein reduces cell viability through induction of cell cycle arrest and apoptosis and, importantly, that a functional p53 tumor suppressor protein is required for these effects. This reduction in cell viability is accompanied by modulation of the levels of p21, p27, and cyclin D1 and by modulation of Akt and PKC pathways. Altogether, our results demonstrate an antitumoral role of HO-1 and points to the importance of p53 status in this antitumor activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
51280
Hemin, BioXtra, from Porcine, ≥97.0% (HPLC)
C34H32ClFeN4O4
H9039
Hemin, from bovine, ≥90%
C34H32ClFeN4O4