EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
BrettPhos ligand supported palladium-catalyzed C-O bond formation through an electronic pathway of reductive elimination: fluoroalkoxylation of activated aryl halides.
PMID 25236851
Abstract
We report an unprecedented BrettPhos ligand supported Pd-catalyzed CO bond-forming reaction of activated aryl halides with primary fluoroalkyl alcohols. We demonstrate that the Phosphine ligand (BrettPhos) possesses the property of altering the mechanistic pathway of reductive elimination from nucleophile to nucleophile. The Pd/BrettPhos catalyst system facilitates the reductive elimination of the oxygen nucleophile through an electronic pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
718742
BrettPhos, 96%
C35H53O2P