EMAIL THIS PAGE TO A FRIEND
Oncotarget
TAp73 promotes cell survival upon genotoxic stress by inhibiting p53 activity.
PMID 25237903
Abstract
p53 plays a key role in regulating DNA damage response by suppressing cell cycle progression or inducing apoptosis depending on extent of DNA damage. However, it is not clear why mild genotoxic stress favors growth arrest, whereas excessive lesions signal cells to die. Here we showed that TAp73, a p53 homologue thought to have a similar function as p53, restrains the transcriptional activity of p53 and prevents excessive activation of its downstream targets upon low levels of DNA damage, which results in cell cycle arrest. Extensive DNA damage triggers TAp73 depletion through ubiquitin/proteasome-mediated degradation of E2F1, leading to enhanced transcriptional activation by p53 and subsequent induction of apoptosis. These findings provide novel insights into the regulation of p53 function and suggest that TAp73 keeps p53 activity in check in regulating cell fate decisions upon genotoxic stress.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6627
5-Fluorouracil, ≥99% (HPLC), powder
C4H3FN2O2
03738
5-Fluorouracil, analytical standard
C4H3FN2O2
858471
5-Fluorouracil, 99%
C4H3FN2O2
C0662
6-Carboxyfluorescein, ~97% (HPLC)
C21H12O7
06-598
Anti-acetyl-Histone H4 Antibody, Anti-acetyl-Histone H4 Antibody is a Rabbit Polyclonal Antibody for detection of acetyl-Histone H4 also known as H4 histone family member A, histone 1-H4a & has been validated in ChIP, ICC, IP & WB.
479306
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
H6Cl2N2Pt
P4394
cis-Diammineplatinum(II) dichloride, crystalline
H6Cl2N2Pt
C2210000
Cisplatin, European Pharmacopoeia (EP) Reference Standard
H6Cl2N2Pt
Y0001018
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
H6Cl2N2Pt
E1383
Etoposide, synthetic, ≥98%, powder
C29H32O13
E2600000
Etoposide, European Pharmacopoeia (EP) Reference Standard
C29H32O13
Y0001365
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
C29H32O13
F8423
Fluorouracil, meets USP testing specifications
C4H3FN2O2
PHR1227
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
C4H3FN2O2
F0250000
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
C4H3FN2O2
1279000
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
C4H3FN2O2
37095
Staurosporine, VETRANAL, analytical standard
C28H26N4O3
P1525
trans-Platinum(II)diammine dichloride
H6Cl2N2Pt
1672803
Transplatin, United States Pharmacopeia (USP) Reference Standard
H6Cl2N2Pt