EMAIL THIS PAGE TO A FRIEND
Oncotarget
IL-6-stimulated CD11b+ CD14+ HLA-DR- myeloid-derived suppressor cells, are associated with progression and poor prognosis in squamous cell carcinoma of the esophagus.
PMID 25238263
Abstract
The aim of this study was to assess the significance of myeloid-derived suppressor cells (MDSCs) and their association with IL-6 in esophageal squamous cell carcinoma (SCC). We examined the percentage of CD11b+CD14+HLA-DR- myeloid cells and the levels of IL-6 in the peripheral blood of 50 patients with esophageal SCC and 12 healthy controls. Moreover, we evaluated the relationship between MDSC recruitment, IL-6 levels, and tumor progression by adding 4-nitroquinoline 1-oxide (4-NQO) to the drinking water of mice to induce esophageal tumors. Here we demonstrated that circulating CD11b+CD14+HLA-DR- cells were significantly increased in esophageal SCC patients compared with healthy people, and this was associated with the clinical stage, treatment response and circulating IL-6 levels. In a 4-NQO-induced esophageal tumor animal model, MDSC recruitment was associated with invasive esophageal tumors and with increased IL-6 levels. IL-6 stimulated reactive oxygen species, arginase 1 and p-STAT3 in MDSCs. Blockade of IL-6 prevented induction of MDSCs and the incidence of 4-NQO- induced invasive tumors. In conclusion, the levels of MDSCs and IL-6 predicted the prognosis of patients with esophageal SCC. Moreover, we suggest inhibition of IL-6 as a potential strategy for the treatment of esophageal SCC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N8141
4-Nitroquinoline N-oxide, ≥98%
C9H6N2O3
442683
4-Nitroquinoline N-oxide, analytical standard
C9H6N2O3