EMAIL THIS PAGE TO A FRIEND
Applied and Environmental Microbiology
Cloning and Characterization of a Novel Esterase from Rhodococcus sp. for Highly Enantioselective Synthesis of a Chiral Cilastatin Precursor.
PMID 25239898