EMAIL THIS PAGE TO A FRIEND
Eukaryotic Cell
A high-throughput fatty acid profiling screen reveals novel variations in fatty acid biosynthesis in Chlamydomonas reinhardtii and related algae.
PMID 25239975
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T80489
Trimethylsulfonium iodide, 98%
C3H9IS