EMAIL THIS PAGE TO A FRIEND
Journal of Pharmaceutical and Biomedical Analysis
Capillary electromigration techniques as tools for assessing the status of vitamins A, C and E in patients with cystic fibrosis.
PMID 25240158
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
296988
Octane, anhydrous, ≥99%
C8H18
657042
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
C8H18
74820
Octane, analytical standard
C8H18
47773
Phylloquinone (K1), analytical standard
C31H46O2
P1609000
Phytomenadione, European Pharmacopoeia (EP) Reference Standard
C31H46O2
1538006
Phytonadione, United States Pharmacopeia (USP) Reference Standard
C31H46O2
PHR1078
Phytonadione, Pharmaceutical Secondary Standard; Certified Reference Material
C31H46O2
17772
Retinol, ≥95.0% (HPLC), ~2700 U/mg
C20H30O
229946
Sodium tetraborate, 99.998% trace metals basis
B4Na2O7
221732
Sodium tetraborate, 99%
B4Na2O7
T4625
Thiamine hydrochloride, reagent grade, ≥99% (HPLC)
C12H17ClN4OS · HCl
T1270
Thiamine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
C12H17ClN4OS · HCl
51622
Thiamine hydrochloride, PharmaGrade, EP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
C12H17ClN4OS · HCl
PHR1037
Thiamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C12H17ClN4OS · HCl
1656002
Thiamine hydrochloride, United States Pharmacopeia (USP) Reference Standard
C12H17ClN4OS · HCl
04573
Thiamine hydrochloride, tested according to Ph.Eur.
C12H17ClN4OS · HCl
T4562
Thiamine hydrochloride, meets USP testing specifications
C12H17ClN4OS · HCl
W332208
Thiamine hydrochloride, ≥98%, FCC, FG
C12H17ClN4OS · HCl
Y0000467
Thiamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C12H17ClN4OS · HCl
47858
Thiamine Hydrochloride (B1), analytical standard
C12H17ClN4OS · HCl
V3501
Vitamin K1, viscous liquid
C31H46O2
95271
Vitamin K1, BioXtra, ≥99.0% (sum of isomers, HPLC), mixtur of isomers
C31H46O2