EMAIL THIS PAGE TO A FRIEND
Journal of Dermatological Science
Retinoic acid and hydroquinone induce inverse expression patterns on cornified envelope-associated proteins: implication in skin irritation.
PMID 25240866