EMAIL THIS PAGE TO A FRIEND
Analytical and Bioanalytical Chemistry
Analysis of whole blood samples with low gas flow inductively coupled plasma-optical emission spectrometry.
PMID 25240935
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
86944
Hexamethyldisilazane, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
C6H19NSi2