EMAIL THIS PAGE TO A FRIEND
The Chinese journal of physiology
Effects of anti-histamine treatment on liver injury triggered by small intestinal ischemia reperfusion in rats.
PMID 25241987
Abstract
Mast cell (MC) degranulation has been implicated in small intestinal ischemia reperfusion (IIR) injury, therein, inhibiting overproduction of histamine released from activated MC may provide promising strategies against IIR-mediated liver injuries. The aim of the present study was to explore whether anti-histamine treatment contribute to attenuating IIR-mediated liver injury. Adult SD rats were randomized into sham-operated group (S group), sole IIR group (IIR group), and IIR treated with Ketotifen, a histamine antagonist (IIR+K group), Cromolyn Sodium, a MC stabilizer (IIR+C group), and Compound 48/80, a MC degranulator (IIR+CP group), respectively. IIR was induced by superior mesenteric artery occlusion for 75 min followed by 4 h of reperfusion. The agents were intravenously administrated 5 min before reperfusion to induce different levels of histamine. Subsequently, serum concentrations of ALT, AST and histamine; levels of LDH,TNF-α, IL-8 and MDA as well as SOD activities in the liver were assessed. Histopathologic changes were also evaluated. IIR resulted in severe liver injury as demonstrated by significant increases in injury scores, with concomitant significant increases in serum ALT, AST and histamine levels, as well as LDH, TNF-α, IL-8, and MDA levels in the liver, accompanied by reduction in SOD activities (all P < 0.05, IIR vs. S). Treatments by Ketotifen and Cromolyn Sodium similarly markedly alleviated IIR-mediated liver injury as confirmed by significant reduction of the above biomedical changes whereas Compound 48/80 further aggravated IIR-mediated liver injury by dramatically enhancing the above biomedical changes. Data of our study suggest that anti-histamine treatments may provide promising benefits in alleviating liver injury triggered by IIR.