EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
A novel ion-exclusion chromatography-mass spectrometry method to measure concentrations and cycling rates of carbohydrates and amino sugars in freshwaters.
PMID 25242224
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09684
Ammonia, puriss., anhydrous, ≥99.9%
H3N
48260
D-(+)-Galactose, for microbiology, ≥99.0%
C6H12O6
ARK2181
D-(+)-Mannose, PharmaGrade, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
C6H12O6
M2069
D-(+)-Mannose, from wood, ≥99%
C6H12O6
M8574
D-(+)-Mannose, synthetic, ≥99%
C6H12O6
M6020
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
C6H12O6
63579
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
C6H12O6
63580
D-(+)-Mannose, for microbiology, ≥99%
C6H12O6
63582
D-(+)-Mannose, for microbiology, ≥99.0% (sum of enantiomers, HPLC)
C6H12O6
M4625
D-(+)-Mannose
C6H12O6
W379301
D-(−)-Ribose, ≥98%
C5H10O5
R1757
D-(−)-Ribose, 98%
C5H10O5
R7500
D-(−)-Ribose, ≥99%
C5H10O5
R9629
D-(−)-Ribose, suitable for cell culture, BioReagent
C5H10O5
R9633
D-(−)-Ribose, plant cell culture tested, BioReagent
C5H10O5
83860
D-(−)-Ribose, ≥99.0% (sum of enantiomers, HPLC)
C5H10O5
W360600
D-(+)-Xylose, ≥99%
C5H10O5
X1500
D-(+)-Xylose, ≥99%
C5H10O5
X3877
D-(+)-Xylose, BioXtra, ≥99%
C5H10O5
95729
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
C5H10O5
47253
D-(+)-Xylose, analytical standard
C5H10O5
F0127
D-(−)-Fructose, ≥99%
C6H12O6
F2543
D-(−)-Fructose, BioXtra
C6H12O6
F3510
D-(−)-Fructose, BioReagent, suitable for cell culture, suitable for insect cell culture
C6H12O6
F9048
D-(−)-Fructose, meets USP testing specifications
C6H12O6
F2793
D-(−)-Fructose, analytical standard, analytical standard for fructose assay kit, for use with enzymatic assay kit FA20
C6H12O6
47739
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
C6H12O6
47740
D-(−)-Fructose, for microbiology, ≥99.0%
C6H12O6
47748
D-(−)-Fructose, tested according to Ph.Eur.
C6H12O6
414565
Divinylbenzene, technical grade, 80%
C10H10