EMAIL THIS PAGE TO A FRIEND
International immunopharmacology
Protective effects of esculentic acid against endotoxic shock in Kunming mice.
PMID 25242384
Abstract
Esculentic acid (EA), a triterpene compound extracted from the root of Phytolacca esculenta (the Chinese name Shang Lu), has been widely used to therapy a variety of inflammatory diseases such as rheumatoid arthritis, edema, hepatitis and bronchitis. The present study was designed to investigate the protective effects of EA against LPS-induced endotoxic shock by the intraperitoneal injection of EA (1, 5 and 10 mg/kg) prior to LPS stimulation (1 mg/kg, i.p.). We examined the effects of EA on the survival rate of mice, inflammatory cytokine and pro-inflammatory mediator production, histopathological changes and protein expression of COX-2 in tissue sections from lung, liver and kidney. The results indicate that EA not only increases the survival rate of mice, but decreases the levels of TNF-α, IL-6, NO and PGE2 in serum or tissues, histopathological changes and COX-2 protein expression also. Furthermore, EA also increases the levels of anti-inflammatory cytokine IL-10 in serum. Overall, these data suggest that the protective effects of EA against LPS-induced endotoxic shock may be mediated, at least in part, by regulation the release of inflammatory cytokines and mediators, and protein expression of COX-2 in mice.