EMAIL THIS PAGE TO A FRIEND
International journal of systematic and evolutionary microbiology
Hymenobacter roseus sp. nov., isolated from sand.
PMID 25242537
Abstract
Strain JC245(T) was isolated from a sand sample, and appeared as dark pink colonies on agar plates with cells staining Gram-negative. Catalase and oxidase activities were positive. Casein was hydrolysed while chitin, gelatin and starch were not. Major (>5 %) fatty acids were iso-C15 : 0, iso-C15 : 0 3-OH, C16 : 1ω5c, C16 : 1ω6c/C16 : 1ω7c, anteiso-C17 : 1 B/iso-C17 : 1 I and iso-C17 : 0 3-OH. Strain JC245(T) contained phosphatidylethanolamine and two unidentified lipids as the major polar lipids, with minor amounts of four unidentified lipids and an unidentified amino lipid. Bacterial hopane derivatives and adenosylhopane were the major hopanoids. Hydroxyflexixanthin was identified as one of the major carotenoids of strain JC245(T) along with five unidentified carotenoids. The genomic DNA G+C content was 52.5 mol%. 16S rRNA gene sequence comparisons indicated that strain JC245(T) represents a member of the genus Hymenobacter within the family Cytophagaceae of the phylum Bacteroidetes. Strain JC245(T) shared the highest 16S rRNA gene sequence similarity with Hymenobacter roseosalivarius AA-718(T) (98.3 %) and other members of the genus Hymenobacter (<95.1 %). However, strain JC245(T) showed 21±2 % relatedness (based on DNA-DNA hybridization) with H. roseosalivarius DSM 11622(T). Distinct morphological, physiological and genotypic differences from previously described taxa support the classification of strain JC245(T) as a representative of a novel species in the genus Hymenobacter, for which the name Hymenobacter roseus sp. nov. is proposed. The type strain is JC245(T) ( = KCTC 42090(T) = LMG 28260(T)).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90656
17α(H),21β(H)-Hopane solution, 0.1 mg/mL in isooctane, analytical standard
C30H52
07562
17β(H),21β(H)-Hopane solution, 0.1 mg/mL in isooctane, analytical standard
C30H52
60660 3-sn-Phosphatidylethanolamine solution, from soybean, ≥97.0% (10 mg phospholipid per ml CHCl3, TLC)
P7943 L-α-Phosphatidylethanolamine from egg yolk, Type III, ≥97% (TLC), lyophilized powder
P6386 L-α-Phosphatidylethanolamine from egg yolk, Type III, 10 mg/mL in chloroform, ≥97%, solution
P8193 L-α-Phosphatidylethanolamine from Glycine max (soybean), Type IV, ≥98% (TLC), lyophilized powder
420875
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
O2Si
420832
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
O2Si
420840
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420883
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
O2Si
420891
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
O2Si
420824
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
O2Si
420816
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420808
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
O2Si
420794
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
O2Si
420786
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420778
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
O2Si
420859
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
O2Si
748161
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
O2Si
718483
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
O2Si
421553
Silica
O2Si
643645
Silica, mesostructured, MCM-41 type (hexagonal)
O2Si
560979
Silica, mesostructured, MSU-F (cellular foam)
O2Si
541036
Silica, mesostructured, HMS (wormhole)
O2Si
637238
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
O2Si
637246
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
O2Si