EMAIL THIS PAGE TO A FRIEND
The Journal of Pharmacy and Pharmacology
In-situ forming gel-like depot of a polyaspartamide-polylactide copolymer for once a week administration of sulpiride.
PMID 25243336