EMAIL THIS PAGE TO A FRIEND
Microbiology and Immunology
Detection of measles, mumps and rubella viruses by immuno-colorimetric assay and its application in focus reduction neutralization tests.
PMID 25244651