EMAIL THIS PAGE TO A FRIEND
Chronobiology International
Constant light suppresses production of Met-enkephalin-containing peptides in cultured splenic macrophages and impairs primary immune response in rats.
PMID 25245012