EMAIL THIS PAGE TO A FRIEND
Annals of the Royal College of Surgeons of England
Adrenaline hip joint infiltration before portal placement can be used to diminish intra-articular bleeding and improve arthroscopic view during further capsulotomy.
PMID 25245747