EMAIL THIS PAGE TO A FRIEND
Organic letters
Selective synthesis of α,α-dideuterio alcohols by the reduction of carboxylic acids using SmI2 and D2O as deuterium source under SET conditions.
PMID 25247236
Abstract
The first general method for the chemoselective synthesis of α,α-dideuterio alcohols directly from feedstock carboxylic acids under single electron transfer conditions using SmI2 is reported. This reaction proceeds after the activation of Sm(II) with a Lewis base, results in excellent levels of deuterium incorporation across a wide range of substrates, and represents an attractive alternative to processes mediated by pyrophoric alkali metal deuterides.