EMAIL THIS PAGE TO A FRIEND
Anesthesiology
Lamiophlomis rotata, an orally available Tibetan herbal painkiller, specifically reduces pain hypersensitivity states through the activation of spinal glucagon-like peptide-1 receptors.
PMID 25247855