EMAIL THIS PAGE TO A FRIEND
Protoplasma
Modulation of liver and kidney toxicity by herb Withania somnifera for silver nanoparticles: a novel approach for harmonizing between safety and use of nanoparticles.
PMID 25248758