EMAIL THIS PAGE TO A FRIEND
Journal of microbiology and biotechnology
Whole-blood gene-expression profiles of cows infected with Mycobacterium avium subsp. paratuberculosis reveal changes in immune response and lipid metabolism.
PMID 25248984