EMAIL THIS PAGE TO A FRIEND
Journal of Laboratory Automation
Label-free measurements of reaction kinetics using a droplet-based optofluidic device.
PMID 25249275
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56226
Rhodamine 6G, analytical standard
C28H30N2O3 · HCl