EMAIL THIS PAGE TO A FRIEND
Lung cancer (Amsterdam, Netherlands)
A phase II study of erlotinib monotherapy in pre-treated non-small cell lung cancer without EGFR gene mutation who have never/light smoking history: re-evaluation of EGFR gene status (NEJ006/TCOG0903).
PMID 25249428
Abstract
Epidermal growth factor receptor (EGFR) tyrosine kinase inhibitors are particularly effective in non-small cell lung cancer (NSCLC) patients harboring active EGFR mutations. However, some studies have reported survival benefits in NSCLC patients with wild-type EGFR upon erlotinib treatment. This trial was conducted to evaluate the efficacy of erlotinib monotherapy and investigate the predictive values of several biomarkers. Patients with previously treated NSCLC but without EGFR gene mutations that had never or light smoked were eligible for this study. Gene status screening was performed using the PNA-LNA PCR clamp method. Erlotinib was administered until disease progression or unacceptable toxicities occurred. EGFR gene status was re-evaluated using the fragment method to detect exon 19 deletions and the Cycleave-PCR method to detect point mutations. Expression of hepatocyte growth factor (HGF), Met, and thymidylate synthase (TS) were evaluated using immunohistochemistry. Forty-seven patients were enrolled in the study between March 2010 and November 2011. Objective response rate (ORR) and disease control rate (DCR) were 15.2% and 41.3%. Re-evaluations for EGFR gene were performed in 32 tumor samples. EGFR gene mutations were found in eight samples (5:exon 19 deletion, 2:G719X, 1:L858R). Six patients had PR and two had SD among these eight patients. A total of 24 patients were confirmed as wild-type EGFR using different methods. ORR and DCR were 4.2% and 41.7%. The median progression free survival (PFS) and median survival times were 2.0 and 6.0 months, respectively. Patients with tumors expressing HGF showed shorter PFS but not MET or TS. Re-examination of EGFR gene status using different detecting method or different sample should be considered to grasp a chance of erlotinib treatment after first line treatment. In confirmed EGFR wild NSCLC, negative HGF staining could be a biomarker for longer PFS by erlotonib treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2500182
Anti-CACNA1C antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1303610
ANTI-CYP5A1 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
H0652 Anti-Hepatocyte Growth Factor antibody produced in goat, IgG fraction of antiserum, lyophilized powder
H7157 Anti-Hepatocyte Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
H0540 Anti-Hepatocyte Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
H9786 Anti-Hepatocyte Growth Factor Receptor (c-Met) antibody produced in goat, affinity isolated antibody, lyophilized powder
H9911 Anti-Hepatocyte Growth Factor Receptor antibody produced in goat, lyophilized powder, affinity isolated antibody
SAB2101033
Anti-HGF (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2101034
Anti-HGF (ab2) antibody produced in rabbit, affinity isolated antibody
HPA040360
Anti-HGF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2105571
Anti-HGF antibody produced in rabbit, affinity isolated antibody
SAB1410624
Anti-HGF antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA044088
Anti-HGF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300599
Anti-MET (Ab-1003) antibody produced in rabbit, affinity isolated antibody
SAB4300479
Anti-MET (Ab-1234) antibody produced in rabbit, affinity isolated antibody
SAB4300487
Anti-MET (Ab-1349) antibody produced in rabbit, affinity isolated antibody
SAB1406117
Anti-MET antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4501870
Anti-MET antibody produced in rabbit, affinity isolated antibody
HPA031259
Anti-TBXAS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031257
Anti-TBXAS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031258
Anti-TBXAS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1406564
Anti-TYMS antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1411373
Anti-TYMS antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
H1896 Monoclonal Anti-Hepatocyte Growth Factor antibody produced in mouse, clone 24612.111, purified immunoglobulin, lyophilized powder
H0665 Monoclonal Anti-Hepatocyte Growth Factor antibody produced in mouse, clone 24516, purified immunoglobulin, lyophilized powder
WH0007298M1
Monoclonal Anti-TYMS antibody produced in mouse, clone 3A1, purified immunoglobulin, buffered aqueous solution
SAB4100588
Monoclonal Anti-TYMS antibody produced in mouse, clone TYMS-A563, culture supernatant
SAB1404503
Monoclonal Anti-TYMS antibody produced in mouse, clone 2B2, purified immunoglobulin, buffered aqueous solution
AV48543
Anti-TYMS antibody produced in rabbit, affinity isolated antibody