PloS one
Xanthene derivatives increase glucose utilization through activation of LKB1-dependent AMP-activated protein kinase.
PMID 25250787
Abstract
5' AMP-activated protein kinase (AMPK) is a highly conserved serine-threonine kinase that regulates energy expenditure by activating catabolic metabolism and suppressing anabolic pathways to increase cellular energy levels. Therefore AMPK activators are considered to be drug targets for treatment of metabolic diseases such as diabetes mellitus. To identify novel AMPK activators, we screened xanthene derivatives. We determined that the AMPK activators 9H-xanthene-9-carboxylic acid {2,2,2-trichloro-1-[3-(3-nitro-phenyl)-thioureido]-ethyl}-amide (Xn) and 9H-xanthene-9-carboxylic acid {2,2,2-trichloro-1-[3-(3-cyano-phenyl)-thioureido]-ethyl}-amide (Xc) elevated glucose uptake in L6 myotubes by stimulating translocation of glucose transporter type 4 (GLUT4). Treatment with the chemical AMPK inhibitor compound C and infection with dominant-negative AMPKa2-virus inhibited AMPK phosphorylation and glucose uptake in myotubes induced by either Xn or Xc. Of the two major upstream kinases of AMPK, we found that Xn and Xc showed LKB1 dependency by knockdown of STK11, an ortholog of human LKB1. Single intravenous administration of Xn and Xc to high-fat diet-induced diabetic mice stimulated AMPK phosphorylation of skeletal muscle and improved glucose tolerance. Taken together, these results suggest that Xn and Xc regulate glucose homeostasis through LKB1-dependent AMPK activation and that the compounds are potential candidate drugs for the treatment of type 2 diabetes mellitus.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-181
Anti-AMPK α-pan Antibody, Anti-AMPK α-pan Antibody is an antibody against AMPK α-pan for use in WB.
05-724
Anti-Myc Tag Antibody, clone 4A6, This Anti-Myc Tag Antibody, clone 4A6 is validated for use in ChIP, IC, IF, IP, WB for the detection of Myc Tag. This antibody has been cited in more than 25 published papers.
07-303
Anti-phospho-Acetyl CoA Carboxylase (Ser79) Antibody, Anti-phospho-Acetyl CoA Carboxylase (Ser79) Antibody is an antibody against phospho-Acetyl CoA Carboxylase (Ser79) for use in WB.
L7917 Anti-LKB1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB3500366
Anti-LKB1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4502889
Anti-LKB1 antibody produced in rabbit, affinity isolated antibody
SAB4502890
Anti-LKB1 antibody produced in rabbit, affinity isolated antibody
SAB1406494
Anti-STK11 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA017254
Anti-STK11 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV41743
Anti-STK11 antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
EHU106061 MISSION® esiRNA, esiRNA human RPL6 (esiRNA1)
EHU156191 MISSION® esiRNA, esiRNA human TM4SF1 (esiRNA1)
EMU065311 MISSION® esiRNA, esiRNA targeting mouse Tm4sf1 (esiRNA1)
EMU159881 MISSION® esiRNA, esiRNA targeting mouse Rpl6
L9168
Monoclonal Anti-LKB1 antibody produced in mouse, ~1 mg/mL, clone Ley37D/G6, purified immunoglobulin, buffered aqueous solution
WH0006794M1 Monoclonal Anti-STK11 antibody produced in mouse, clone 2A7-H2, ascites fluid