EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
X-pep, a novel cell-penetrating peptide motif derived from the hepatitis B virus.
PMID 25251474
Abstract
Cell-penetrating peptides (CPPs) are able to penetrate the plasma membrane and gain access to the interior of any replicating or non-replicating cell, and are being considered as drug delivery agents. Here we describe the serendipitous discovery of a novel CPP motif (MAARLCCQ), designated X-pep, located at the extreme N-terminus of the X-protein of the hepatitis B virus. X-pep, and a C-terminally truncated form of the peptide (MAARL), readily penetrated HepG2 cells. Further truncation by removal of the terminal leucine residue impaired the cell-penetrating activity of peptide, indicating that MAARL is the active core of the peptide. X-pep is located adjacent to another CPP, namely Xentry, and like Xentry is unable to penetrate unactivated resting lymphocytes suggesting selective cell uptake. A D-isomeric form of the MAARL peptide was not cell-permeable, indicating that the cell-penetrating function of the peptide involves stereoselective interaction with a chiral receptor. The discovery of X-pep, which bears no resemblance to known CPPs, allows studies to be undertaken to determine additional characteristics of this novel CPP.