EMAIL THIS PAGE TO A FRIEND
Journal of water and health
High prevalence of multidrug-resistant Escherichia coli and Enterococcus spp. in river water, upstream and downstream of a wastewater treatment plant.
PMID 25252346
Abstract
In this study, microbial quality and antimicrobial resistance of faecal bacteria from a Portuguese river were assessed. River water samples collected upstream and downstream of a wastewater treatment plant, throughout a 3-month period, were used for the enumeration of Escherichia coli and Enterococcus spp. The highest numbers found for E. coli and enterococci were 1.1 × 10⁴ and 1.2 × 10⁴ colony forming units (CFU)/100 ml, respectively. In total, 144 isolates of E. coli and 144 of enterococci were recovered and tested for antimicrobial susceptibility; 104 E. coli and 78 Enterococcus spp. showed resistance to one or more antimicrobial drugs. Overall, 70 and 32 different resistance patterns were found for E. coli and enterococci, respectively. One E. coli showed resistance to imipenem and 29 isolates were extended spectrum β-lactamase-producers. Multidrug-resistant E. coli belonged mostly to groups A, B1 and group D. Enterococcus spp. were mostly resistant to rifampicin, tetracycline, azithromycin and erythromycin; six isolates showed resistance to vancomycin, presenting the VanA phenotype. The high levels of E. coli and enterococci and the remarkable variety of antimicrobial resistance profiles, reinforces the theory that these river waters can be a pool of antimicrobial resistance determinants, which can be easily spread among different bacteria and reach other environments and hosts.